Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethanolamine

Catalog No. D888525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D888525G 25 g
50-311-9124 500 g
D8885100G 100 g
D88851KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. D888525G Supplier Sigma Aldrich Supplier No. D888525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethanolamine

Specifications

CAS 1080-32-6
Density 1.095 g/mL at 25°C
Boiling Point 106°C to 108°C (1 mmHg, lit.)
Molecular Formula C11H17O3P
Refractive Index n20/D 1.477
Linear Formula C6H5CH2P(O)(OC2H5)2
MDL Number MFCD00009078
Quantity 25 g
Synonym 2,2'-Iminodiethanol; Bis(2-hydroxyethyl)amine
Molecular Weight (g/mol) 228.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.