Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethoxy(3-glycidyloxypropyl)methylsilane

Catalog No. 43517125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-517-125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-517-125ML Supplier Sigma Aldrich Supplier No. 43517125ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethoxy(3-glycidyloxypropyl)methylsilane (GPMS) is an epoxysilane which can be used as a silane coupling agent for the surface treatment of a variety of materials. These silanes can also be used as adhesion promoters by modifying the surface properties of the substrates and elastomeric materials.GPMS can be used in the surface modification of a variety of particles such as cellulose nanocrystals, silica nanomaterials and other silicon based substrates. It functionalizes these surfaces by attaching the epoxy-silane groups with the surface molecules.

Specifications

CAS 2897-60-1
Density 0.978 g/mL (at 25°C)
Boiling Point 122°C to 126°C (5 mmHg)
Molecular Formula C11H24O4Si
Refractive Index n20/D 1.431 (literature)
Linear Formula C11H24O4Si
Quantity 25 mL
MDL Number MFCD00046995
Synonym (3-Glycidyloxypropyl)methyldiethoxysilane; [3-(2,3-Epoxypropoxy)propyl]methyldiethoxysilane
Molecular Weight (g/mol) 248.39
Percent Purity 97%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.