Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethoxydimethylsilane

Catalog No. 175595500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-559-5500ML 500 mL
17-559-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-559-5500ML Supplier Sigma Aldrich Supplier No. 175595500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethoxydimethylsilane (DEDMS) is an organically modified silicate (ORMOSIL) with two methyl groups in each unit that facilitate the hydrophobic characteristics. It is mainly utilized to functionalize a variety of substrates.The composite of DEDMS and triethoxymethylsilane can be used to synthesize a liquid precursor resin for the preparation of silicon oxycarbide (SiOC). It can form a sol-gel coating on polyamide-polyethylene (PA/PE), which can be used as an anti-oxidizing food packaging material. It can also be used as a precursor in the synthesis of polydimethylsiloxane, which forms an anti-corrosive coating composite with waterborne polyurethane.

Specifications

CAS 78-62-6
Density 0.865 g/mL (at 25°C)
Boiling Point 114°C
Molecular Formula C6H16O2Si
Refractive Index n20/D 1.381 (literature)
Linear Formula (CH3)2 Si(OC2H5)2
Quantity 500 mL
MDL Number MFCD00009068
Synonym Dimethyldiethoxysilane
Molecular Weight (g/mol) 148.28
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.