Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethoxydimethylsilane, 97%

Catalog No. AC174632500
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC174632500 250 mL
AC174631000 100 mL
AC174635000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC174632500 Supplier Thermo Scientific Chemicals Supplier No. 174632500

CAS: 78-62-6 | C6H16O2Si | 148.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-62-6
MDL Number MFCD00009068
InChI Key YYLGKUPAFFKGRQ-UHFFFAOYSA-N
Synonym dimethyldiethoxysilane, silane, diethoxydimethyl, diethoxy dimethyl silane, dimethyl-diethoxysilan, dimethylsilicondiethoxide, dimethyl diethoxy silane, unii-i3931j3zj2, dimethyl-diethoxysilan czech, ccris 1321, kbe 22
PubChem CID 62322
IUPAC Name diethoxy(dimethyl)silane
SMILES CCO[Si](C)(C)OCC

Specifications

Color Colorless
Density 0.86g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C6H16O2Si
Refractive Index 1.3805 to 1.3825 (20°C, 589nm)
Linear Formula (CH3)2Si(OC2H5)2
Quantity 250 mL
Fieser 13,76
Molecular Weight (g/mol) 148.277
Formula Weight 148.28
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Diethoxydimethylsilane, 97%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.