Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Diethoxydiphenylsilane, 98%
Click to view available options
Quantity:
10g
50g
250g
Specifications
Beilstein | 2281302 |
Formula Weight | 272.42 |
Percent Purity | 98% |
Density | 1.033 |
Refractive Index | 1.5255 |
Sensitivity | Moisture sensitive |
Boiling Point | 166°C to 168°C (15mmHg) |
Quantity | 50g |
Flash Point | 175°C (347°F) |
Chemical Name or Material | Diethoxydiphenylsilane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 219-860-5
TSCA : Yes
Recommended Storage : Ambient temperatures