Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl 1,4-cyclohexanedione-2,5-dicarboxylate

Catalog No. 12612810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-612-810G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-612-810G Supplier Sigma Aldrich Supplier No. 12612810G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethyl 1,4-cyclohexanedione-2,5-dicarboxylate

Specifications

CAS 787-07-5
Melting Point 127°C to 129°C (lit.)
Molecular Formula C12H16O6
Linear Formula (O=)2C6H6(CO2C2H5)2
MDL Number MFCD00001608
Quantity 100 mg
Synonym Diethyl succinylosuccinate
Molecular Weight (g/mol) 256.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.