Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate | 632-93-9 | MFCD00005950 | 99.8% | 267.32 g/mol | C14H21NO4 | 25 G
SDP

Catalog No. 5000229540
Encompass_Preferred

Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate is a small-molecule research reagent used in biochemical studies and reported as a ferrochelatase inhibitor. It is supplied as a solid or as a solution in DMSO and is intended for laboratory research applications.

  • High purity: 99.8%.
  • Molecular weight: 267.32 g/mol.
  • Chemical formula: C14H21NO4.
  • Available as solid or DMSO solution.
  • Commonly used in studies of heme metabolism and ferrochelatase activity.

Catalog No. 50-002-29540 Supplier Medchemexpress LLC Supplier No. HY11859425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate is a small-molecule research reagent used in biochemical studies and reported as a ferrochelatase inhibitor. It is supplied as a solid or as a solution in DMSO and is intended for laboratory research applications.

  • High purity: 99.8%.
  • Molecular weight: 267.32 g/mol.
  • Chemical formula: C14H21NO4.
  • Available as solid or DMSO solution.
  • Commonly used in studies of heme metabolism and ferrochelatase activity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.