Learn More
Medchemexpress LLC Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate | 1149-23-1 | MFCD00005951 | 253.30 | C13H19NO4 | 250 G
SDP
Catalog No. 5000302343
Encompass_Preferred
Diludine (Et-HE) is Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate (CAS 1149-23-1), a small-molecule antioxidant that modulates oxidative-stress pathways including superoxide dismutase (SOD). It is used in research on inflammation and oxidative stress and has reported applications in improving antioxidative status during the dairy cow transition period.
  • Antioxidant activity targeting oxidative stress and SOD pathway.
  • Chemical formula C13H19NO4 and molecular weight 253.30.
  • Used in studies of immunology, inflammation, and oxidative-stress biology.
  • Reported application: improving antioxidative status in dairy cows during transition.
  • Supplied in bulk laboratory pack sizes, including 250 g, 500 g, and 1 kg.
Catalog No. 50-003-02343 Supplier Medchemexpress LLC Supplier No. 250GHYW017960
Diludine (Et-HE) is Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate (CAS 1149-23-1), a small-molecule antioxidant that modulates oxidative-stress pathways including superoxide dismutase (SOD). It is used in research on inflammation and oxidative stress and has reported applications in improving antioxidative status during the dairy cow transition period.
  • Antioxidant activity targeting oxidative stress and SOD pathway.
  • Chemical formula C13H19NO4 and molecular weight 253.30.
  • Used in studies of immunology, inflammation, and oxidative-stress biology.
  • Reported application: improving antioxidative status in dairy cows during transition.
  • Supplied in bulk laboratory pack sizes, including 250 g, 500 g, and 1 kg.