Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 1,4-Phenylenediacetate 98.0+%, TCI America™
SDP

Catalog No. P14345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14345G 5 g
P143425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14345G Supplier TCI America Supplier No. P14345G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36076-26-3
Molecular Formula C14H18O4
Molecular Weight (g/mol) 250.294
MDL Number MFCD00032366
InChI Key JRSOAUOHTHOAOP-UHFFFAOYSA-N
Synonym 1,4-Phenylenediacetic Acid Diethyl Ester
PubChem CID 284070
IUPAC Name ethyl 2-[4-(2-ethoxy-2-oxoethyl)phenyl]acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)CC(=O)OCC

Specifications

Melting Point 59°C
Color White-Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 250.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 1,4-Phenylenediacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.