Learn More
Cayman Chemical Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate, Purity: ≥98%, CAS: 632-93-9, 25 g
SDP
Catalog No. 502964117
Encompass_Preferred
An inhibitor of heme production; inhibits ferrochelatase; induces Mallory-Denk body formation in mouse liver; reduces IL-12A methylation in mouse liver
Catalog No. 50-296-4117 Supplier Cayman Chemical Supplier No. 2323025 G
An inhibitor of heme production; inhibits ferrochelatase; induces Mallory-Denk body formation in mouse liver; reduces IL-12A methylation in mouse liver