Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 2,2':6',2″-Terpyridine-4'-phosphonate 93.0+%, TCI America™
SDP

Catalog No. D4511200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4511200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4511200MG Supplier TCI America Supplier No. D4511200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 161583-75-1
Molecular Formula C19H20N3O3P
Molecular Weight (g/mol) 369.361
MDL Number MFCD11042551
InChI Key MDIMWBGOUZAXEP-UHFFFAOYSA-N
Synonym 2,2′C:6′C,2′C′C-Terpyridine-4′C-phosphonic Acid Diethyl Ester
PubChem CID 71307775
IUPAC Name 4-diethoxyphosphoryl-2,6-dipyridin-2-ylpyridine
SMILES CCOP(=O)(C1=CC(=NC(=C1)C2=CC=CC=N2)C3=CC=CC=N3)OCC

Specifications

Melting Point 149°C
Color Yellow
Quantity 200 mg
Formula Weight 369.36
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 2,2′:6′,2′′-Terpyridine-4′-phosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.