Learn More
Medchemexpress LLC Diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate | 632-93-9 | MFCD00005950 | 99.5% | 267.32 g/mol | C14H21NO4 | 10 G
SDP
Catalog No. 5000226300
Encompass_Preferred
Diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate is a diethyl ester of a 1,4-dihydropyridine used as a small-molecule reagent or synthetic intermediate in research laboratories.
  • White to off-white solid suitable for laboratory use.
  • Molecular formula C14H21NO4; molecular weight 267.32 g/mol.
  • High purity (99.5% assay) appropriate for analytical and synthetic work.
  • Stable as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • In solvent recommended storage -80°C (6 months) or -20°C (1 month).
  • Available in multiple sizes, including 10 G, for flexibility in experiments.
  • Used as a dihydropyridine ester intermediate or reagent in chemical research.
Catalog No. 50-002-26300 Supplier Medchemexpress LLC Supplier No. HY11859410G
Diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate is a diethyl ester of a 1,4-dihydropyridine used as a small-molecule reagent or synthetic intermediate in research laboratories.
  • White to off-white solid suitable for laboratory use.
  • Molecular formula C14H21NO4; molecular weight 267.32 g/mol.
  • High purity (99.5% assay) appropriate for analytical and synthetic work.
  • Stable as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • In solvent recommended storage -80°C (6 months) or -20°C (1 month).
  • Available in multiple sizes, including 10 G, for flexibility in experiments.
  • Used as a dihydropyridine ester intermediate or reagent in chemical research.