Learn More
Medchemexpress LLC Diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate | 632-93-9 | MFCD00005950 | 99.8% | 267.32 g·mol-1 | C14H21NO4 | 500 MG
SDP
Catalog No. 5000226314
Encompass_Preferred
Diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate is a synthetic dihydropyridine derivative supplied as a white to off-white powder for research use. It is used as a reference standard in biochemical and pharmacological studies and is reported to inhibit ferrochelatase, interfering with heme synthesis pathways.
  • High purity suitable for analytical and research applications.
  • Molecular weight 267.32 g·mol-1 and chemical formula C14H21NO4.
  • White to off-white solid supplied as a powder.
  • Store powder at -20°C for long-term or 4°C for short-term; store in solvent at -80°C for extended stability.
  • Common applications include biochemical assays and studies of heme biosynthesis.
Catalog No. 50-002-26314 Supplier Medchemexpress LLC Supplier No. HY118594500MG
Diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate is a synthetic dihydropyridine derivative supplied as a white to off-white powder for research use. It is used as a reference standard in biochemical and pharmacological studies and is reported to inhibit ferrochelatase, interfering with heme synthesis pathways.
  • High purity suitable for analytical and research applications.
  • Molecular weight 267.32 g·mol-1 and chemical formula C14H21NO4.
  • White to off-white solid supplied as a powder.
  • Store powder at -20°C for long-term or 4°C for short-term; store in solvent at -80°C for extended stability.
  • Common applications include biochemical assays and studies of heme biosynthesis.