Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 2,5-Dibromoterephthalate 98.0+%, TCI America™
SDP

Catalog No. D39955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39955G 5 g
D399525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39955G Supplier TCI America Supplier No. D39955G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18013-97-3
Molecular Formula C12H12Br2O4
Molecular Weight (g/mol) 380.032
MDL Number MFCD12407163
InChI Key WXRSDHICEYICMV-UHFFFAOYSA-N
Synonym 2,5-Dibromoterephthalic Acid Diethyl Ester
PubChem CID 22146229
IUPAC Name diethyl 2,5-dibromobenzene-1,4-dicarboxylate
SMILES CCOC(=O)C1=CC(=C(C=C1Br)C(=O)OCC)Br

Specifications

Melting Point 129°C
Color White
Boiling Point 335°C
Quantity 5 g
Formula Weight 380.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 2,5-Dibromoterephthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.