Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 2-Ethyl-2-methylmalonate 97.0+%, TCI America™
SDP

Catalog No. D36305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36305G 5 g
D363025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36305G Supplier TCI America Supplier No. D36305G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2049-70-9
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD02258541
InChI Key ODRGILDUWDVBJX-UHFFFAOYSA-N
Synonym 2-Ethyl-2-methylmalonic Acid Diethyl Ester
PubChem CID 16293
IUPAC Name 1,3-diethyl 2-ethyl-2-methylpropanedioate
SMILES CCOC(=O)C(C)(CC)C(=O)OCC

Specifications

Color Yellow
Boiling Point 79°C
Quantity 5 g
Formula Weight 202.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 2-Ethyl-2-methylmalonate
EINECSNumber (2)-0861&(2)-0879
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.