Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl (2-Oxo-2-Phenylethyl)Phosphonate

Catalog No. 41075610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-075-610G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-075-610G Supplier Sigma Aldrich Supplier No. 41075610G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant involved in:Asymmetric Michael addition of +a-oxo phosphonates to nitro olefinsGem-chlorofluorination of keto phosphonates with subsequent functionalization of the productsCyclocondensation reactions to produce arylphosphonatesDiazo transfer reactions for synthesis of diazo-phosphonyl compoundsHorner-Wadsworth-Emmons reactionsInverse-electron-demand Diels-Alder reactions

Specifications

CAS 31410-07-8
Molecular Formula C7H11BrN2
Refractive Index n20/D 1.513
Linear Formula C7H11BrN2
MDL Number MFCD08277003
Quantity 5 g
Synonym Diethyl benzoylmethylphosphonate; Diethyl phenacylphosphonate; NSC 648426
Molecular Weight (g/mol) 203.08 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.