Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 2-Propynylmalonate 97.0+%, TCI America™
SDP

Catalog No. D46165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46165G 5 g
D46161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46165G Supplier TCI America Supplier No. D46165G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17920-23-9
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00091967
InChI Key FKXOELIKVUIBLP-UHFFFAOYSA-N
Synonym 2-Propynylmalonic Acid Diethyl Ester, Diethyl Propargylmalonate, Propargylmalonic Acid Diethyl Ester
PubChem CID 28835
IUPAC Name diethyl 2-prop-2-ynylpropanedioate
SMILES CCOC(=O)C(CC#C)C(=O)OCC

Specifications

Color Colorless
Boiling Point 115°C
Quantity 5 g
Formula Weight 198.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 2-Propynylmalonate
RTECSNumber OO1630000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.