Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl(3-Bromopropyl)Phosphonate

Catalog No. 5420325ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-203-25ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 54-203-25ML Supplier Sigma Aldrich Supplier No. 5420325ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for synthesis of: Hybrid assembly composed of titanium dioxide nanoparticles and thin multi-walled carbon nanotubesOrganosoluble zirconium phosphonate nanocomposites for asymmetric hydrogenationCyclic hydroxamic acids via alkylation followed by intramolecular cyclizationpH-responsive cyclopolymerReactant for: Homogenization of zirconium hydroxide phosphonate supported ruthenium catalystReactions with sodium imidazolide

Specifications

CAS 21176-60-3
Refractive Index n20/D 1.4620
Linear Formula C6H5CH2CH2CH(NH2)CO2H -+ HCl
MDL Number MFCD03427064
Quantity 1 g
Synonym 1,2-Diaminoethane trityl polystyrene resin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.