Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diethyl 3-oxopentanedioate | 105-50-0 | ≥97.0% | 202.21 g·mol⁻¹ | C9H14O5 | 50 G
SDP

Catalog No. 5000302719
Encompass_Preferred

Diethyl 3-oxopentanedioate (diethyl acetonedicarboxylate) is an organic biochemical reagent used as a building block in organic synthesis and in biochemical assay development. It is supplied for laboratory research applications.

  • High purity (≥97.0%).
  • Molecular formula C9H14O5; molecular weight 202.21 g·mol⁻¹.
  • Suitable for organic synthesis and biochemical assay development.
  • Available in multiple package sizes, including 50 g.
  • Solutions available (10 mM in DMSO, 1 mL) for screening workflows.

Catalog No. 50-003-02719 Supplier Medchemexpress LLC Supplier No. HYW01479950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethyl 3-oxopentanedioate (diethyl acetonedicarboxylate) is an organic biochemical reagent used as a building block in organic synthesis and in biochemical assay development. It is supplied for laboratory research applications.

  • High purity (≥97.0%).
  • Molecular formula C9H14O5; molecular weight 202.21 g·mol⁻¹.
  • Suitable for organic synthesis and biochemical assay development.
  • Available in multiple package sizes, including 50 g.
  • Solutions available (10 mM in DMSO, 1 mL) for screening workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.