Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diethyl 3-oxopentanedioate | 105-50-0 | MFCD00009200 | 97.0% | 202.21 g/mol | C9H14O5 | 100 G
SDP

Catalog No. 5000303600
Encompass_Preferred

Diethyl 3-oxopentanedioate (CAS 105-50-0) is a biochemical reagent used in life-science research and organic synthesis. It is supplied both as solid material and as ready-to-use DMSO solutions for assay preparation, and is intended for laboratory research use only.

  • Purity 97.0% (typical).
  • Molecular formula C9H14O5; molecular weight 202.21 g/mol.
  • Available as DMSO solutions and solid packs (25 g-1 kg) for flexible use.
  • SDS and certificate of analysis available for handling and quality verification.
  • Suitable as a synthetic building block and biochemical assay reagent.

Catalog No. 50-003-03600 Supplier Medchemexpress LLC Supplier No. HYW014799100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethyl 3-oxopentanedioate (CAS 105-50-0) is a biochemical reagent used in life-science research and organic synthesis. It is supplied both as solid material and as ready-to-use DMSO solutions for assay preparation, and is intended for laboratory research use only.

  • Purity 97.0% (typical).
  • Molecular formula C9H14O5; molecular weight 202.21 g/mol.
  • Available as DMSO solutions and solid packs (25 g-1 kg) for flexible use.
  • SDS and certificate of analysis available for handling and quality verification.
  • Suitable as a synthetic building block and biochemical assay reagent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.