Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diethyl 3-oxopentanedioate | 105-50-0 | MFCD00009200 | ≥97.0% | 202.21 g·mol⁻¹ | C9H14O5 | 25 G
SDP

Catalog No. 5000303010
Encompass_Preferred

Diethyl 3-oxopentanedioate is an organic biochemical reagent used in life-science research and organic synthesis. It appears as a colorless to light-yellow liquid with defined purity and storage recommendations and is intended for research use only.

  • High purity (≥97.0%).
  • Molecular weight 202.21 g·mol⁻1.
  • Chemical formula C9H14O5.
  • Colorless to light-yellow liquid; density 1.113 g·cm⁻3.
  • Available in multiple pack sizes, including 25 g.
  • Suitable for biochemical assays and synthetic applications.
  • Recommended storage: -20°C for long-term stability.

Catalog No. 50-003-03010 Supplier Medchemexpress LLC Supplier No. 25GHYW014799
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethyl 3-oxopentanedioate is an organic biochemical reagent used in life-science research and organic synthesis. It appears as a colorless to light-yellow liquid with defined purity and storage recommendations and is intended for research use only.

  • High purity (≥97.0%).
  • Molecular weight 202.21 g·mol⁻1.
  • Chemical formula C9H14O5.
  • Colorless to light-yellow liquid; density 1.113 g·cm⁻3.
  • Available in multiple pack sizes, including 25 g.
  • Suitable for biochemical assays and synthetic applications.
  • Recommended storage: -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.