Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl (5-Bromopentyl)malonate 97.0+%, TCI America™
SDP

Catalog No. B20835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20835G 5 g
B208325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20835G Supplier TCI America Supplier No. B20835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1906-95-2
Molecular Formula C12H21BrO4
Molecular Weight (g/mol) 309.2
MDL Number MFCD00048565
InChI Key JZCAZGWJCVMTTQ-UHFFFAOYSA-N
Synonym (5-Bromopentyl)malonic Acid Diethyl Ester
PubChem CID 4401294
IUPAC Name diethyl 2-(5-bromopentyl)propanedioate
SMILES CCOC(=O)C(CCCCCBr)C(=O)OCC

Specifications

Boiling Point 161°C
Quantity 5 g
Formula Weight 309.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (5-Bromopentyl)malonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.