Learn More
Apexbio Technology LLC Diethyl Acetamidomalonate 1068-90-2 500g
SDP
Catalog No. 5000095012
Encompass_Preferred
Diethyl Acetamidomalonate (1068-90-2) is an organic compound containing both amide and ester functional groups It is primarily utilized in chemical synthesis for producing amino acid derivatives and other biologically active molecules Diethyl Acetamidomalonate enables the introduction of acetylamino groups into target compounds via synthetic transformations such as nucleophilic substitution Based on these properties Diethyl Acetamidomalonate holds research potential in the development of novel amino acid analogues and bioactive small-molecule compounds supporting studies in medicinal chemistry and pharmaceutical development
Catalog No. 50-000-95012 Supplier Apexbio Technology LLC Supplier No. A7053500000
Diethyl Acetamidomalonate (1068-90-2) is an organic compound containing both amide and ester functional groups It is primarily utilized in chemical synthesis for producing amino acid derivatives and other biologically active molecules Diethyl Acetamidomalonate enables the introduction of acetylamino groups into target compounds via synthetic transformations such as nucleophilic substitution Based on these properties Diethyl Acetamidomalonate holds research potential in the development of novel amino acid analogues and bioactive small-molecule compounds supporting studies in medicinal chemistry and pharmaceutical development