Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl adipate

Catalog No. 245720100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-572-0100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 24-572-0100G Supplier Sigma Aldrich Supplier No. 245720100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethyl adipate

Specifications

CAS 141-28-6
Melting Point -20°C to +19°C
Density 1.009 g/mL (at 25°C)
Boiling Point 251°C
Molecular Formula C10H18O4
Refractive Index n20/D 1.427 (literature)
Linear Formula C2H5OCO(CH2)4 COOC2H5
Quantity 100 g
MDL Number MFCD00009215
Synonym Hexanedioic acid 1,6-diethyl ester; NSC 19160; NSC 3363
Molecular Weight (g/mol) 202.25
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.