Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl Allylphosphonate

Catalog No. 5654155G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-541-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 56-541-55G Supplier Sigma Aldrich Supplier No. 5654155G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for: Copolymerization of phosphonated allyl monomers and maleic anhydrideEnantioselective synthesis of solamin type mono-THF acetogeninsRCM reaction yielding oxaphospholene and oxaphosphinene heterocyclesSynthesis of spongistatin 2 using Wittig couplingStereoselective synthesis of pentacyclic furanosteroidsPreparation of protected polyhydroxylated +a -amino acid constitutents of microsclerodermins

Specifications

CAS 544-97-8
Density 0.724 g/mL at 25°C
Boiling Point 44°C to 46°C
Molecular Formula C2H6Zn
Refractive Index n20/D 1.4340
Linear Formula (CH3)2Zn
Quantity 5 g
MDL Number MFCD00014854
Synonym 2-Propenyl-phosphonic acid diethyl ester
Molecular Weight (g/mol) 95.46 g/mol
Biological Activity Central Nervous System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.