Learn More
Medchemexpress LLC Diethyl aminoethyl hexanoate citrate | 220439-24-7 | 99.9% | 407.46 | 1 ML
SDP
Catalog No. 5000435928
Encompass_Preferred
Diethyl aminoethyl hexanoate citrate is a compound widely used as a plant growth regulator. It stimulates the regeneration of adventitious buds and can effectively cause Cd extraction, fixing more Cd in cell walls and reducing Cd migration in shoots to reduce metal toxicity. When combined with gibberellic acid 3 and EDTA, it alleviates the adverse effect of EDTA on plant growth.
  • Used as a plant growth regulator
  • Stimulates regeneration of adventitious buds
  • Causes Cd extraction, fixes more Cd in cell walls, and reduces Cd migration in shoot to reduce metal toxicity
  • Alleviates adverse effect of EDTA on plant growth when combined with gibberellic acid 3 and EDTA
Catalog No. 50-004-35928 Supplier Medchemexpress LLC Supplier No. HY112106A10MM/1ML
Diethyl aminoethyl hexanoate citrate is a compound widely used as a plant growth regulator. It stimulates the regeneration of adventitious buds and can effectively cause Cd extraction, fixing more Cd in cell walls and reducing Cd migration in shoots to reduce metal toxicity. When combined with gibberellic acid 3 and EDTA, it alleviates the adverse effect of EDTA on plant growth.
  • Used as a plant growth regulator
  • Stimulates regeneration of adventitious buds
  • Causes Cd extraction, fixes more Cd in cell walls, and reduces Cd migration in shoot to reduce metal toxicity
  • Alleviates adverse effect of EDTA on plant growth when combined with gibberellic acid 3 and EDTA