Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl aminomalonate hydrochloride

Catalog No. D87589100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D87589100G 100 g
D8758925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D87589100G Supplier Sigma Aldrich Supplier No. D87589100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethyl aminomalonate hydrochloride

Specifications

CAS 13433-00-6
Melting Point 165°C to 170°C (dec.) (lit.)
Molecular Formula C7H13NO4 -+ HCl
Linear Formula NH2CH2(COOC2H5)2 -+ HCl
MDL Number MFCD00012510
Quantity 100 g
Synonym Aminomalonic acid diethyl ester hydrochloride
Molecular Weight (g/mol) 211.64
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.