Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Benzylidenemalonate 96.0+%, TCI America™
SDP

Catalog No. B042025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B042025G 25 g
B04205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B042025G Supplier TCI America Supplier No. B042025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5292-53-5
Molecular Formula C14H16O4
Molecular Weight (g/mol) 248.28
MDL Number MFCD00009149
InChI Key VUWPIBNKJSEYIN-UHFFFAOYSA-N
Synonym diethyl benzylidenemalonate, diethyl benzalmalonate, diethyl 2-benzylidenemalonate, ethyl benzylidenemalonate, diethylbenzalmalonate, diethyl phenylmethylene malonate, malonic acid, benzylidene-, diethyl ester, diethylmalonate, benzal, benzylidenemalonic acid diethyl ester, propanedioic acid, phenylmethylene-, diethyl ester
PubChem CID 94751
IUPAC Name 1,3-diethyl 2-(phenylmethylidene)propanedioate
SMILES CCOC(=O)C(=CC1=CC=CC=C1)C(=O)OCC

Specifications

Color Yellow
Quantity 25 g
Formula Weight 248.28
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Benzylidenemalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.