Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl Benzylphosphonate

Catalog No. D91071100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D91071100G 100 g
D9107125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D91071100G Supplier Sigma Aldrich Supplier No. D91071100G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for synthesis of: 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disordersNatural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactionsStilbenes via on-column oxidation of vicinal diols and Horner-Emmons reactionsInhibitors of teh Wnt pathway for colon cancer repression using Wadsworth-Emmons reactionsAntimalarial drug analogs against P. falciparumReactant for:Cyclization of aryl ethers, amines, and amidesInvestigating the effects of functional groups on the performance of clue organic LEDs

Specifications

CAS 5324-30-1
Density 1.348 g/mL at 25°C
Boiling Point 75°C (1 mmHg, lit.)
Molecular Formula C6H14BrO3P
Refractive Index n20/D 1.497
Linear Formula BrCH2CH2P(O)(OCH2CH3)2
MDL Number MFCD00000239
Quantity 100 g
Synonym (Diethoxyphosphonomethyl)benzene; Benzylphosphonic acid diethyl ester; NSC 62294
Molecular Weight (g/mol) 245.05 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.