Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl Benzylphosphonate

Catalog No. D9107125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D9107125G 25 g
D91071100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D9107125G Supplier Sigma Aldrich Supplier No. D9107125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for synthesis of: 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disordersNatural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactionsStilbenes via on-column oxidation of vicinal diols and Horner-Emmons reactionsInhibitors of teh Wnt pathway for colon cancer repression using Wadsworth-Emmons reactionsAntimalarial drug analogs against P. falciparumReactant for:Cyclization of aryl ethers, amines, and amidesInvestigating the effects of functional groups on the performance of clue organic LEDs

Specifications

CAS 1080-32-6
Density 1.095 g/mL at 25°C
Boiling Point 106°C to 108°C (1 mmHg, lit.)
Molecular Formula C11H17O3P
Refractive Index n20/D 1.497
Linear Formula C6H5CH2P(O)(OC2H5)2
MDL Number MFCD00009078
Quantity 25 g
Synonym (Diethoxyphosphonomethyl)benzene; Benzylphosphonic acid diethyl ester; NSC 62294
Molecular Weight (g/mol) 228.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.