Learn More
Medchemexpress LLC Diethyl bipy55 DC 1g | 1762-46-5 | 1 G
SDP
Catalog No. 5000228978
Encompass_Preferred
Diethyl 2,2'-bipyridine-5,5'-dicarboxylate (CAS 1762-46-5) is the diethyl ester of 2,2'-bipyridine-5,5'-dicarboxylic acid. It is used as a ligand precursor and building block in coordination chemistry and organic synthesis, supplied as a small-scale research pack for laboratory use.
  • Diethyl ester derivative of 2,2'-bipyridine dicarboxylic acid.
  • Commonly used as a ligand precursor in coordination chemistry.
  • Useful building block for organic synthesis and catalysis studies.
  • Supplied as a 1 G pack suitable for laboratory-scale research.
  • Typical reported purity ≥98%; confirm certificate of analysis before use.
Catalog No. 50-002-28978 Supplier Medchemexpress LLC Supplier No. HY1168931G
Diethyl 2,2'-bipyridine-5,5'-dicarboxylate (CAS 1762-46-5) is the diethyl ester of 2,2'-bipyridine-5,5'-dicarboxylic acid. It is used as a ligand precursor and building block in coordination chemistry and organic synthesis, supplied as a small-scale research pack for laboratory use.
  • Diethyl ester derivative of 2,2'-bipyridine dicarboxylic acid.
  • Commonly used as a ligand precursor in coordination chemistry.
  • Useful building block for organic synthesis and catalysis studies.
  • Supplied as a 1 G pack suitable for laboratory-scale research.
  • Typical reported purity ≥98%; confirm certificate of analysis before use.