Learn More
Medchemexpress LLC Diethyl bipy55 DC 500mg | 1762-46-5 | 300.31 g/mol | C16H16N2O4 | 500 MG
SDP
Catalog No. 5000228233
Encompass_Preferred
Diethyl [2,2'-bipyridine]-5,5'-dicarboxylate is a research-grade bipyridine derivative used as a ligand in coordination chemistry and as an inhibitor in biochemical studies. It is supplied as a high-purity solid for synthesis and assay applications.
  • High purity: 98.79% (HPLC).
  • Molecular weight 300.31 g/mol; formula C16H16N2O4.
  • Effective ligand for coordination chemistry and organometallic synthesis.
  • Used in biochemical assays as a CP4H inhibitor.
  • Supplied as a solid powder with COA and SDS available.
Catalog No. 50-002-28233 Supplier Medchemexpress LLC Supplier No. HY116893500MG
Diethyl [2,2'-bipyridine]-5,5'-dicarboxylate is a research-grade bipyridine derivative used as a ligand in coordination chemistry and as an inhibitor in biochemical studies. It is supplied as a high-purity solid for synthesis and assay applications.
  • High purity: 98.79% (HPLC).
  • Molecular weight 300.31 g/mol; formula C16H16N2O4.
  • Effective ligand for coordination chemistry and organometallic synthesis.
  • Used in biochemical assays as a CP4H inhibitor.
  • Supplied as a solid powder with COA and SDS available.