Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl (Bromodifluoromethyl)Phosphonate

Catalog No. 4113615G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-136-15G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-136-15G Supplier Sigma Aldrich Supplier No. 4113615G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat3)Synthesis of Mycobacterium tuberculosis protein tyrosine phosphatase inhibitory difluoromethylphosphonic acid derivatives via multi step synthesis with Suzuki coupling and resolution as key stepsP-C bond cleavage on basic hydrolysisPreparation of a prodrug for intracellular delivery of protein tyrosine phosphatase 1B (PTP1B) inhibitor (nonhydrolyzable difluoromethylphosphonate moiety)

Specifications

CAS 1095-03-0
Melting Point 98°C to 101°C
Molecular Formula C18H15BO3
Refractive Index n20/D 1.417
Linear Formula (C6H5O)3B
MDL Number MFCD00059011
Quantity 10 g
Synonym (Bromodifluoromethyl)phosphonic acid diethyl ester
Molecular Weight (g/mol) 290.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.