Learn More
Medchemexpress LLC Diethyl butylmalonate | 133-08-4 | MFCD00009170 | >99.0% | 216.27 g/mol | C11H20O4 | 10 MG
SDP
Catalog No. 5000299223
Encompass_Preferred
Diethyl butylmalonate is a malonate ester used as a research reagent that acts as a succinate dehydrogenase (SDH) inhibitor and modulates reactive oxygen species (ROS) in biochemical studies. It is provided for laboratory research use only and is not for human or clinical applications.
  • Succinate dehydrogenase inhibitor commonly used in SDH/ROS studies.
  • Chemical formula C11H20O4 and molecular weight 216.27 g/mol.
  • Available in solution formats and multi-gram solid packages from manufacturers.
  • Intended for research use only; consult safety data sheet for handling.
Catalog No. 50-002-99223 Supplier Medchemexpress LLC Supplier No. HY44178S10MG
Diethyl butylmalonate is a malonate ester used as a research reagent that acts as a succinate dehydrogenase (SDH) inhibitor and modulates reactive oxygen species (ROS) in biochemical studies. It is provided for laboratory research use only and is not for human or clinical applications.
  • Succinate dehydrogenase inhibitor commonly used in SDH/ROS studies.
  • Chemical formula C11H20O4 and molecular weight 216.27 g/mol.
  • Available in solution formats and multi-gram solid packages from manufacturers.
  • Intended for research use only; consult safety data sheet for handling.