Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl carbamazine citrate salt, MP Biomedicals™

Catalog No. ICN19019225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19019225 25 g
ICN19019210 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19019225 Supplier MP Biomedicals, Inc Supplier No. 0219019225

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1642-54-2
Molecular Formula C16H29N3O8
Molecular Weight (g/mol) 391.42
MDL Number MFCD00039124
InChI Key PGNKBEARDDELNB-UHFFFAOYSA-N
Synonym diethylcarbamazine citrate, loxuran, caritrol, dicarocide, eosinopin, filazine, franocide, franozan, carbam palatabs, ethodryl citrate
PubChem CID 15432
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid; N,N-diethyl-4-methylpiperazine-1-carboxamide
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)C(=O)N1CCN(C)CC1

Specifications

Color White
Quantity 25 g
Formula Weight 391.4
Physical Form Powder
Chemical Name or Material Diethyl carbamazine citrate salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.