Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl carbonate, 99%

Catalog No. AAA124770F
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA124770F 2500 mL
AAA12477AP 500 mL
AAA124770D 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA124770F Supplier Thermo Scientific Chemicals Supplier No. A124770F
  • $165.10
    $132.65 / Each of 1

    Save $32.45

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 105-58-8 | C5H10O3 | 118.13 g/mol

Diethyl carbonate is esters, beta-enamino esters, carbamates and unsymmetrical alkyl carbonates. It is an active component of electrolytes used in lithium which is used as a solvent for cellulose ethers, nitro cellulose, natural and synthetic resin and in erythromycin intramuscular injections. It is also used in the synthesis of 3-ethyl-4-methyl-5-phenyl-3H-oxazol-2-one, phenobarbital, pyrethrum batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl carbonate is esters, beta-enamino esters, carbamates and unsymmetrical alkyl carbonates. It is an active component of electrolytes used in lithium which is used as a solvent for cellulose ethers, nitro cellulose, natural and synthetic resin and in erythromycin intramuscular injections. It is also used in the synthesis of 3-ethyl-4-methyl-5-phenyl-3H-oxazol-2-one, phenobarbital, pyrethrum batteries.

Solubility
Miscible with diehtyl ether, ethanol and chloroform.Immiscible with water.

Notes
Store in cool place. Moisture sensitive. Incompatible with strong acids, oxidizing agents, strong bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-58-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00009107
InChI Key OIFBSDVPJOWBCH-UHFFFAOYSA-N
Synonym ethyl carbonate, carbonic acid, diethyl ester, carbonic acid diethyl ester, eufin, diatol, diaethylcarbonat, carbonic ether, ethoxyformic anhydride, diethylkarbonat, ethyl carbonate eto 2co
PubChem CID 7766
IUPAC Name diethyl carbonate
SMILES CCOC(=O)OCC

Specifications

Melting Point -43°C
Density 0.975
Boiling Point 126°C to 128°C
Flash Point 31°C (87°F)
Odor Pleasant
Assay Percent Range ≥99%
Refractive Index 1.3845
Quantity 2500 mL
UN Number UN2366
Beilstein 956591
Sensitivity Moisture sensitive
Merck Index 14,3780
Solubility Information Miscible with diehtyl ether,ethanol and chloroform.Immiscible with water.
Formula Weight 118.13
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Diethyl carbonate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIETHYL CARBONATE
EINECSNumber 203-311-1
RTECSNumber FF9800000
TSCA Yes
storageNote1 Moisture Sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.