Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl chlorophosphate

Catalog No. D9163225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D9163225G 25 g
D91632100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D9163225G Supplier Sigma Aldrich Supplier No. D9163225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethyl chlorophosphate is a general reagent to synthesize diethylphosphate esters from alcohols, as in the synthesis of organophosphorus nerve agent mimics, chiral phosphine complexes for preparing copper alkoxide phosphine catalysts and the synthesis of chalcogenorhodamine bearing phosphonic acid sensitizers.

Specifications

CAS 814-49-3
Density 1.194 g/mL (at 25°C (literature))
Boiling Point 60°C (2 mmHg)
Molecular Formula C4H10ClO3P
Refractive Index n20/D 1.416 (literature)
Linear Formula (C2H5O)2 P(O)Cl
MDL Number MFCD00009075
Quantity 25 g
Synonym Diethyl phosphorochloridate
Molecular Weight (g/mol) 172.55
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.