Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl Cyanomethylphosphonate

Catalog No. D9170550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D9170550G 50 g
D9170510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D9170550G Supplier Sigma Aldrich Supplier No. D9170550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethyl Cyanomethylphosphonate

Specifications

CAS 1470-61-7
Melting Point 172°C to 175°C
Molecular Formula C5H10AgNS2
Refractive Index n20/D 1.434
Linear Formula (C2H5)2NCSSAg
MDL Number MFCD00004929
Quantity 50 g
Synonym (Diethylphosphono)acetonitrile; Cyanomethylphosphonic acid diethyl ester
Molecular Weight (g/mol) 256.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.