Alfa Aesar™ Diethyl cyanomethylphosphonate, 98+%

Manufacturer: Alfa Aesar™ A1021814

Quantity 25g

 View more versions of this product

Catalog No. AAA1021814

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 2537-48-6
    Molecular Formula C6H12NO3P
    Formula Weight 177.14
    MDL Number MFCD00001893
    UN Number UN3278
    Beilstein 1772981
    Synonym Cyanomethylphosphonic acid diethyl ester; Diethyl phosphonoacetonitrile
    Chemical Name or Material Diethyl cyanomethylphosphonate

    Specifications

    Formula Weight 177.14
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 1772981
    Health Hazard 1 Danger
    Molecular Formula C6H12NO3P
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    MDL Number MFCD00001893
    CAS 2537-48-6
    UN Number UN3278
    Boiling Point 132° to 135°C (1.2mmHg)
    Flash Point >110°C (230°F)
    Synonym Cyanomethylphosphonic acid diethyl ester; Diethyl phosphonoacetonitrile
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
    Quantity 25g
    Chemical Name or Material Diethyl cyanomethylphosphonate
    Assay ≥98%

    Description

    Applications
    Diethyl cyanomethyl phosphonate is used as an intermediate in Horner-Emmons reaction for the synthesis of substituted nitriles and their amide and heterocyclic derivatives. It is known as a modified Wittig reagent used in the preparation of alpha, beta-unsaturated nitriles from ketones or aldehydes like 3-hydroxy-3-methylbutanal. It reacts with epoxides and nitrones to prepare cyano-substituted cyclopropanes and aziridines respectively. It is actively involved in the synthesis of alpha-arylated alkanenitriles via reaction with aryl iodides in presence of CuI.

    Solubility
    Miscible with water, chloroform, terahydrofuran, ethyl acetate and methylene chloride.

    Notes
    Incompatible with acids, bases, oxidizing agents and reducing agents.

    GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit