Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl cyanomethylphosphonate, 96%

Catalog No. p-7080573
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1021814 25 g
AAA1021822 100 g
AAA1021836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1021814 Supplier Thermo Scientific Chemicals Supplier No. A1021814
Only null left
Add to Cart

CAS: 2537-48-6 | C6H12NO3P | 177.14 g/mol

Diethyl cyanomethyl phosphonate is used as an intermediate in Horner-Emmons reaction for the synthesis of substituted nitriles and their amide and heterocyclic derivatives. It is known as a modified Wittig reagent used in the preparation of alpha, beta-unsaturated nitriles from ketones or aldehydes like 3-hydroxy-3-methylbutanal. It reacts with epoxides and nitrones to prepare cyano-substituted cyclopropanes and aziridines respectively. It is actively involved in the synthesis of alpha-arylated alkanenitriles via reaction with aryl iodides in presence of CuI.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl cyanomethyl phosphonate is used as an intermediate in Horner-Emmons reaction for the synthesis of substituted nitriles and their amide and heterocyclic derivatives. It is known as a modified Wittig reagent used in the preparation of alpha, beta-unsaturated nitriles from ketones or aldehydes like 3-hydroxy-3-methylbutanal. It reacts with epoxides and nitrones to prepare cyano-substituted cyclopropanes and aziridines respectively. It is actively involved in the synthesis of alpha-arylated alkanenitriles via reaction with aryl iodides in presence of CuI.

Solubility
Miscible with water, chloroform, terahydrofuran, ethyl acetate and methylene chloride.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2537-48-6
Molecular Formula C6H12NO3P
Molecular Weight (g/mol) 177.14
MDL Number MFCD00001893
InChI Key KWMBADTWRIGGGG-UHFFFAOYSA-N
Synonym diethyl cyanomethylphosphonate, diethyl cyanomethyl phosphonate, diethylphosphono acetonitrile, cyanomethylphosphonic acid diethyl ester, diethylphosphonoacetonitrile, diethylcyanomethylphosphonate, diethyl cyanomethanephosphonate, phosphonic acid, cyanomethyl-, diethyl ester, cyanomethyl phosphonic acid diethyl ester
PubChem CID 75676
IUPAC Name 2-diethoxyphosphorylacetonitrile
SMILES CCOP(=O)(CC#N)OCC

Specifications

Density 1.094
Boiling Point 132°C to 135°C (1.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.433
Quantity 25 g
UN Number UN3278
Beilstein 1772981
Solubility Information Miscible with water,chloroform,terahydrofuran,ethyl acetate and methylene chloride.
Formula Weight 177.14
Percent Purity ≥98%
Chemical Name or Material Diethyl cyanomethylphosphonate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
EINECSNumber 219-806-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.