Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Diethyl-D-tartarate, MP Biomedicals™

Catalog No. ICN15674101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15674101 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15674101 Supplier MP Biomedicals, Inc Supplier No. 0215674101

missing translation for 'validMainframeMsg'

([-]-Tartaric acid diethyl ester), 1mL = approx. 1.2g

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13811-71-7
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.19
MDL Number MFCD00064451
InChI Key YSAVZVORKRDODB-WDSKDSINSA-N
Synonym --diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
PubChem CID 117410
IUPAC Name 1,4-diethyl (2S,3S)-2,3-dihydroxybutanedioate
SMILES CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC

Specifications

Quantity 1 mL
Formula Weight 206.2
Physical Form Liquid
Chemical Name or Material (-)-Diethyl-D-tartarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.