Learn More
Medchemexpress LLC Diethyl decanedioate | 110-40-7 | 99.77% | 258.36 | 5 G
SDP
Catalog No. 5000366874
Encompass_Preferred
Diethyl decanedioate is an organic compound used as a raw material for flavors and food additives. It can be used to make fruit and mint flavors and is widely applied in food processing and cosmetic production. Additionally, this compound serves as a reagent and intermediate in certain organic synthesis reactions. It is also a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • Used as a raw material for flavors and food additives
  • Can be used to make fruit and mint flavors
  • Widely applied in food processing and cosmetic production
  • Serves as a reagent and intermediate in organic synthesis reactions
  • Biochemical reagent for life science-related research
Catalog No. 50-003-66874 Supplier Medchemexpress LLC Supplier No. HYW0094435G
Diethyl decanedioate is an organic compound used as a raw material for flavors and food additives. It can be used to make fruit and mint flavors and is widely applied in food processing and cosmetic production. Additionally, this compound serves as a reagent and intermediate in certain organic synthesis reactions. It is also a biochemical reagent that can be used as a biological material or organic compound for life science-related research.
  • Used as a raw material for flavors and food additives
  • Can be used to make fruit and mint flavors
  • Widely applied in food processing and cosmetic production
  • Serves as a reagent and intermediate in organic synthesis reactions
  • Biochemical reagent for life science-related research