Learn More
Medchemexpress LLC Diethyl decanedioate | 110-40-7 | 99.8% | 258.36 | 25 G
SDP
Catalog No. 5000451489
Encompass_Preferred
Diethyl decanedioate is an organic compound commonly used as a raw material for flavors and food additives. It can be used to make fruit and mint flavors, and is widely used in some food processing and cosmetic production. Additionally, this compound serves as a reagent and intermediate in some organic synthesis reactions. As a biochemical reagent, it can be utilized as a biological material or organic compound for life science-related research.
  • Used as a raw material for flavors and food additives (fruit and mint flavors)
  • Widely used in food processing and cosmetic production
  • Functions as a reagent and intermediate in organic synthesis reactions
  • Biochemical reagent for life science-related research
Catalog No. 50-004-51489 Supplier Medchemexpress LLC Supplier No. 25GHYW009443
Diethyl decanedioate is an organic compound commonly used as a raw material for flavors and food additives. It can be used to make fruit and mint flavors, and is widely used in some food processing and cosmetic production. Additionally, this compound serves as a reagent and intermediate in some organic synthesis reactions. As a biochemical reagent, it can be utilized as a biological material or organic compound for life science-related research.
  • Used as a raw material for flavors and food additives (fruit and mint flavors)
  • Widely used in food processing and cosmetic production
  • Functions as a reagent and intermediate in organic synthesis reactions
  • Biochemical reagent for life science-related research