Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl difluoromalonate, 97%

Catalog No. AAL1604206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1604206 5 g
AAL1604203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1604206 Supplier Thermo Scientific Chemicals Supplier No. L1604206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 680-65-9 | C7H10F2O4 | 196.15 g/mol

Diethyl difluoromalonate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl difluoromalonate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 680-65-9
Molecular Formula C7H10F2O4
Molecular Weight (g/mol) 196.15
MDL Number MFCD00221617
InChI Key WCRVWLHTROHXBB-UHFFFAOYSA-N
Synonym diethyl 2,2-difluoromalonate, diethyl difluoromalonate, 2,2-difluoro-malonic acid diethyl ester, 1,3-diethyl 2,2-difluoropropanedioate, difluoromalonic acid diethyl ester, diethyl2,2-difluoromalonate, propanedioic acid, 2,2-difluoro-, 1,3-diethyl ester, propanedioic acid, difluoro-, diethyl ester, diethyl 2,2-difluoromalonate #, diethyl2,2-difluoropropanedioatev
PubChem CID 260648
IUPAC Name diethyl 2,2-difluoropropanedioate
SMILES CCOC(=O)C(F)(F)C(=O)OCC

Specifications

Density 1.179
Boiling Point 94°C to 95°C (23 mmHg)
Refractive Index 1.38
Quantity 5 g
Beilstein 1783527
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 196.15
Percent Purity 97%
Chemical Name or Material Diethyl difluoromalonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.