Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl ether-d{10}, 99%(Isotopic)

Catalog No. AA4228203
Change view
Click to view available options
Quantity:
1 g
Packaging:
Prescored Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA4228203 1 g Prescored Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4228203 Supplier Thermo Scientific Chemicals Supplier No. 04228203
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2679-89-2 | C4H10O | 84.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2679-89-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 84.18
MDL Number MFCD00062316
InChI Key RTZKZFJDLAIYFH-MWUKXHIBSA-N
Synonym ether-d10, diethyl ether-d10, 2,2'-oxybis 2h5 ethane, diethylether-d10, di ethyl-d5 ether, ether-d10, 99 atom % d, 2,2/'-oxybis 2h5 ethane, diethyl ether-d isotopic, 1-2 h? ethoxy 2 h? ethane, 1,1,1,2,2-pentadeuterio-2-1,1,2,2,2-pentadeuterioethoxy ethane
PubChem CID 2723765
IUPAC Name 1-[(1,1,2,2,2-²H₅)ethoxy](²H₅)ethane
SMILES [2H]C([2H])([2H])C([2H])([2H])OC([2H])([2H])C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Packaging Prescored Ampule
Quantity 1 g
Density 0.82 g/mL
Flash Point −40°C (−40°F)
Odor Ether-like
Melting Point -116°C
Boiling Point 33°C to 34°C
Linear Formula (C2D5)2O
UN Number UN1155
Formula Weight 84.19
Percent Purity 99%
Chemical Name or Material Diethyl ether-d{10}, Isotopic
Show More Show Less
Hazard Category H224-H302-H336
Hazard Statement GHS H Statement
H224-H302-H336
Extremely flammable liquid and vapor.
Harmful if swallowed.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: DIETHYL ETHER
EINECSNumber 220-235-4
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.