Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Diethyl ether, HPLC Grade, 99%, stab. with ethanol

Catalog No. AA38990M1
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38990M1 4 x 1 L
AA38990K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38990M1 Supplier Thermo Scientific Chemicals Supplier No. 038990M1
  • $332.50
    $266.65 / Case of 4

    Save $65.85

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

Widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents. | CAS: 60-29-7 | C4H10O | 74.12 g/mol

It is a widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl ether
Melting Point °C to 116°C
CAS 60-29-7
Density 0.713 g/mL
Quantity 4 x 1 L
Flash Point -45°C (-49°F)
Physical Form Liquid
Assay Percent Range 99%
Molecular Formula C4H10O
MDL Number MFCD00011646
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
Boiling Point 34.6°C
Solubility Information Immiscible with water,miscible with alcohol.
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES CCOCC
IUPAC Name ethoxyethane
Molecular Weight (g/mol) 74.12
PubChem CID 3283
ChEBI CHEBI:35702
Formula Weight 74.12
Grade HPLC
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.