Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Ether Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019495 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9495 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9495 Supplier MilliporeSigma Supelco Supplier No. PHR16663X1.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diethyl ether is a non-polar volatile organic solvent, commonly used as an extraction solvent in liquid-liquid extraction for separation of post reaction residues from synthetic products.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 60-29-7
Ignition Point 320°F
Vapor Density 2.6 (vs. air)
Density 0.706 g/mL (at 25°C)
Linear Formula (CH3 CH2)2 O
MDL Number MFCD00011646
Melting Point -116°C
Boiling Point 34.6°C
Synonym Diethyl ether; Ether; Ethyl ether
Type Certified Reference Material
Quantity 3 x 1.2 mL
Formula Weight 74.12
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.