Learn More
Apexbio Technology LLC DIETHYL-FUMARATE-500MG
SDP
Catalog No. 5000568591
Encompass_Preferred
Diethyl fumarate (CAS No 623-91-6) is prominently used in biomedical research particularly in studies focusing on its chemical properties and biological activities Originating as an ester of fumaric acid it is known for modulating cellular pathways often targeting oxidative stress and inflammation-related mechanisms Its in vitro activity is observed in the nanomolar to low micromolar range making it a valuable compound in assays evaluating cellular responses Typical applications include its use in various cell models to study antioxidant effects and in animal studies to explore therapeutic potential against inflammatory conditions Diethyl fumarate is also employed in drug screening processes where its ability to influence specific biochemical pathways is of interest Experimental concentrations typically depend on the specific research design allowing for flexibility in its application across numerous study types
Catalog No. 50-005-68591 Supplier Apexbio Technology LLC Supplier No. C6538500
Diethyl fumarate (CAS No 623-91-6) is prominently used in biomedical research particularly in studies focusing on its chemical properties and biological activities Originating as an ester of fumaric acid it is known for modulating cellular pathways often targeting oxidative stress and inflammation-related mechanisms Its in vitro activity is observed in the nanomolar to low micromolar range making it a valuable compound in assays evaluating cellular responses Typical applications include its use in various cell models to study antioxidant effects and in animal studies to explore therapeutic potential against inflammatory conditions Diethyl fumarate is also employed in drug screening processes where its ability to influence specific biochemical pathways is of interest Experimental concentrations typically depend on the specific research design allowing for flexibility in its application across numerous study types