Learn More
Medchemexpress LLC HY-W010056 500mg Medchemexpress, Diethyl fumarate CAS:623-91-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW010056500MG
Encompass_Preferred
Medchemexpress, HY-W010056 500mg Diethyl fumarate CAS:623-91-6 Diethyl fumarate is a decomposition product of Malathion (an insecticide). Diethyl fumarate is a reputed skin irritant. Diethyl fumarate can causes non-immunologic contact urticaria on skin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3279