Learn More
Medchemexpress LLC Diethyl fumarate | 623-91-6 | 99.0% | 172.18 | 10 G
SDP
Catalog No. 5000366923
Encompass_Preferred
Diethyl fumarate is a decomposition product of Malathion 9 and a reactant in the Diels-Alder reaction. It is a reputed skin irritant and can cause non-immunologic contact urticaria on the skin. Diethyl fumarate and Curcumin hybridize to exert neuroprotective effects.
  • Decomposition product of Malathion 9.
  • Reactant in Diels-Alder reaction.
  • Causes dose-dependent swelling of ears in guinea pigs.
  • Curcumin hybrids show neuroprotective effects in C. elegans model of PD.
  • For research use only.
Catalog No. 50-003-66923 Supplier Medchemexpress LLC Supplier No. HYW01005610G
Diethyl fumarate is a decomposition product of Malathion 9 and a reactant in the Diels-Alder reaction. It is a reputed skin irritant and can cause non-immunologic contact urticaria on the skin. Diethyl fumarate and Curcumin hybridize to exert neuroprotective effects.
  • Decomposition product of Malathion 9.
  • Reactant in Diels-Alder reaction.
  • Causes dose-dependent swelling of ears in guinea pigs.
  • Curcumin hybrids show neuroprotective effects in C. elegans model of PD.
  • For research use only.